missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calcitonin Rabbit Monoclonal Antibody

Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Epredia™ Calcitonin Rabbit Monoclonal Antibody.

£141.78 - £663.00

Specifications

Antigen Calcitonin
Clone SP17
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12683747
View Documents Promotion Details
Epredia™
RM-9117-S0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12663747
View Documents Promotion Details
Epredia™
RM-9117-S1
Concentrated 500 μL
£417.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12673747
View Documents Promotion Details
Epredia™
RM-9117-S
Concentrated 1 mL
£663.00
1mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12633747
View Documents Promotion Details
Epredia™
RM-9117-R7
Prediluted 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12653747
View Documents Promotion Details
Epredia™
RM-9117-RQ
Ready-to-Use 12 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.

Host Species: Rabbit

Clone: SP17

Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Epitope: 1-32aa

Immunogen: Synthetic human calcitonin 1-32 amino acid peptide

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Calcitonin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Synthetic human calcitonin 1-32 amino acid peptide
SP17
Monoclonal
Rabbit
Steroids Hormones and Growth Factors
Primary
Videos
SDS
Documents

Documents

Product Certifications