missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin R Antibody (31/01-1H10-4-1-14) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-697
This item is not returnable.
View return policy
Description
Calcitonin R Monoclonal specifically detects Calcitonin R in Human, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Calcitonin R | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| calcitonin receptor, CRT, CTR, CT-R, CTR1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This recognises an epitope within the cytoplasmic domain that is common to both C1a and C1b human isoforms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 31/01-1H10-4-1-14 | |
| Unconjugated | |
| P30988 | |
| CALCR | |
| Synthetic peptide, DIPIYICHQELRNEPANN, derived from the cytoplasmic domain of human calcitonin receptor. | |
| 0.05 mg | |
| Cancer, GPCR, Signal Transduction | |
| 799 | |
| Human, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction