missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcitonin Monoclonal Antibody (SP17)
Description
Recommended positive controls: Thyroid or Medullary carcinoma.
Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.
Specifications
Specifications
| Antigen | Calcitonin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP17 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.09% sodium azide |
| Gene | CALCA |
| Gene Accession No. | P01258, P06881 |
| Gene Alias | Alpha CGRP; Alpha type CGRP; Alpha-type CGRP; CA; Cal1; CAL6; CALC; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide 1; calcitonin gene related peptide I; calcitonin gene related peptide I precursor; Calcitonin gene-related peptide; calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin precursor; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-like; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRPI; CGRP-I; CT; Ctn; katacalcin; katacalcin (KC); KC; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin; RATCAL6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?