missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin Antibody (SP17), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-26483-0.1ML
This item is not returnable.
View return policy
Description
Calcitonin Monoclonal specifically detects Calcitonin in Human, Rat, Canine, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Calcitonin | |
| Monoclonal | |
| Unconjugated | |
| No buffer with 0.05% Sodium Azide | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Rabbit | |
| Tissue culture supernatant | |
| Cancer, Cellular Markers, Neuroscience | |
| 796 | |
| Store at 4C. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SP17 | |
| Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| P01258 | |
| CALCA | |
| Synthetic human calcitonin 1-32 amino acid peptide. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine, Mouse (Negative) | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction