missing translation for 'onlineSavingsMsg'
Learn More

Calcitonin Antibody (CALCA/3310), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18396433
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396433 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396433 Supplier Bio-Techne Supplier No. NBP314041AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calcitonin Monoclonal antibody specifically detects Calcitonin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcitonin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CALCA/3310
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump
Host Species Mouse
Immunogen Recombinant fragment of human Calcitonin protein (around aa 3-116) (exact sequence is proprietary) (Uniprot: P01258 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.