missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Calcitonin (8-32), Salmon
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Specifications
Specifications
| Species | Salmon |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 2725.06 |
| Product Type | Calcitonin (8-32), salmon |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction