missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Calcineurin B Protein

Product Code. 18496780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18496780 0.5 mg 0.50mg
18355671 100μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18496780 Supplier Novus Biologicals™ Supplier No. NBP1787970.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-170 of Human Calcineurin B The Recombinant Human Calcineurin B Protein is derived from E. coli. The Recombinant Human Calcineurin B Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000936
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2 mM DTT, 0.1 M NaCl.
Gene ID (Entrez) 5534
Molecular Weight (g/mol) 21.5 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human PPP3R1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen PPP3R1, 1-170 aa, Human, His tag, E. coli (NP_000936).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Calcineurin B
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.