missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin B Mouse anti-Human, Clone: CALNB/2342, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | Calcineurin B |
|---|---|
| Clone | CALNB/2342 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Calcineurin B Monoclonal specifically detects Calcineurin B in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Calcineurin B | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 5534 | |
| IgG1 κ | |
| Protein A or G purified |
| CALNB/2342 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa) | |
| Recombinant full-length Calcineurin B protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title