missing translation for 'onlineSavingsMsg'
Learn More

Calcineurin B Antibody (CALNB/2342) - Azide and BSA Free, Novus Biologicals™

Product Code. 18382416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382416 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382416 Supplier Novus Biologicals Supplier No. NBP308464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calcineurin B Monoclonal specifically detects Calcineurin B in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin B
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CALNB/2342
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa)
Host Species Mouse
Immunogen Recombinant full-length Calcineurin B protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5534
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.