missing translation for 'onlineSavingsMsg'
Learn More

Calcineurin B Antibody (4E1), Novus Biologicals™

Product Code. 18350429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350429 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350429 Supplier Novus Biologicals Supplier No. H00005534M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calcineurin B Monoclonal antibody specifically detects Calcineurin B in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin B
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4E1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH27913
Gene Alias calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa)
Host Species Mouse
Immunogen PPP3R1 (AAH27913, 1 a.a. ~ 170 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGNEASYPLEMCSHFDADEIKRLGKRFKKLDLDNSGSLSVEEFMSLPELQQNPLVQRVIDIFDTDGNGEVDFKEFIEGVSQFSVKGDKEQKLRFAFRIYDMDKDGYISNGELFQVLKMMVGNNLKDTQLQQIVDKTIINADKDGDGRISFEEFCAVVGGLDIHKKMVVDV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5534
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.