missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Calcineurin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen Calcineurin
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066973
View Documents
Novus Biologicals™
NBP3-30781-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30066894
View Documents
Novus Biologicals™
NBP3-30781-100ug
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcineurin Polyclonal antibody specifically detects Calcineurin in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifications

Specifications

Calcineurin
Polyclonal
Purified
RUO
Phosphate buffered saline
The immunogen used for the anti-Calcineurin was a purified peptide conjugated to KLH. The sequence of the peptide is NH2-Arg-Met-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro-Ser-Asp-Ala-COOH (a.a. 482-494) corresponding to a 60kDa protein
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Bovine, Human, Mouse, Rat
5530
IgG
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.