Learn More
Invitrogen™ Calcineurin A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529209
Description
Recommended positive controls: Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, Molt-4, U87-MG, SK-N-SH, SK-N-AS, IMR32, JC, BCL-1, rat brain, human Calcineurin A-transfected 293T. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (90%), Xenopus laevis (98%), Dog (100%), Pig (99%), Rabbit (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.
Specifications
| Calcineurin A | |
| Polyclonal | |
| Unconjugated | |
| PPP3CA | |
| 2900074D19Rik; Ca2+/calmodulin-regulated protein phosphatase; calcineurin; calcineurin A alpha; Calcineurin A1; calcineurin catalytic subunit; calcineurin catalytic subunit delta isoform; Calcineurin subunit A alpha; CalcineurinA; calmodulin-dependent calcineurin A subunit alpha isoform; caln; CALNA; CALNA 1; CALNA1; CAM-PRP catalytic subunit; CCN 1; ccn1; CN; CnA; CNA alpha; CNA1; PP2B alpha 1; PP2BA alpha; PP2CB; ppp2b; ppp3ca; ppp3ca.S; protein phosphatase; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform (calcineurin A alpha); protein phosphatase 3 catalytic subunit alpha; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme S homeolog; protein phosphatase 3, catalytic subunit, gamma isozyme; protein phosphatase-2Ba; serine/threonine protein kinase; serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform; XELAEV_18009418mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19055, 24674, 5530 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.98 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P63328, P63329, Q08209 | |
| PPP3CA | |
| Recombinant protein encompassing a sequence within the center region of human Calcineurin A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Amphibian, Human, Mouse, Rodent, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.