missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCUN1D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96995-50ul
This item is not returnable.
View return policy
Description
DCUN1D1 Polyclonal antibody specifically detects DCUN1D1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| DCUN1D1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 μL/mg of lysate | |
| DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae), DCN1-like protein 1, DCUN1L1DCUN1 domain-containing protein 1, Defective in cullin neddylation protein 1-like protein 1, RP42 homolog, RP42Squamous cell carcinoma-related oncogene, SCCROSCRO, Tes3 | |
| Produced in rabbits immunized with purified, recombinant Human DCUN1D1 (Accession#: NP_065691.2; Met1-Val259) | |
| 50 μg | |
| Cancer | |
| 54165 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction