missing translation for 'onlineSavingsMsg'
Learn More

Calciferol, 97%, Thermo Scientific Chemicals

Product Code. 15434589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434589 5 g 5g
15424589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434589 Supplier Thermo Scientific Alfa Aesar Supplier No. J62163.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-14-6 | Molecular Formula: C28H44O | Molecular Weight: 396.66 g/mol

Calciferol has a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol has also been shown to be an inhibitor of DNA polymerase. Calciferol with calcium is used to treat or prevent bone loss (osteoporosis). Calciferol is also used with other medications to treat low levels of calcium or phosphate caused by certain disorders (such as hypoparathyroidism, pseudohypoparathyroidism, familial hypophosphatemia). It is often used in kidney disease to keep calcium levels normal and allow normal bone growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol (80 mg/ml at 25°C), water (<1mg/mL at 25°C), methanol, DMF (∼20 mg/ml), and DMSO (20 mg/ml at 25°C).

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Melting Point 118°C to 119°C
Color White
Quantity 5 g
UN Number UN2811
Beilstein 1916682
Merck Index 14,10018
Solubility Information Soluble in ethanol (80mg/ml at 25°C),water (<1mg/ml at 25°C),methanol,DMF (∽20mg/ml),and DMSO (20mg/ml at 25°C).
Optical Rotation +110° (c=1.5 in Ethanol)
Formula Weight 396.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Calciferol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.