missing translation for 'onlineSavingsMsg'
Learn More

Calciferol

Product Code. p-7042185
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15454179

Brand: Thermo Scientific Alfa Aesar J61610.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-14-6 | C28H44O | 396.66 g/mol

Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

Solubility
Soluble in ethanol, water, methanol, dimethylformamide, and dimethyl sulfoxide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Melting Point 114°C to 118°C
Color White
Quantity 1 g
UN Number UN2811
Beilstein 1916682
Merck Index 14,10018
Solubility Information Soluble in ethanol,water,methanol,dimethylformamide,and dimethyl sulfoxide.
Formula Weight 396.65
Physical Form Powder
Chemical Name or Material Calciferol
compliance-icons compliance-icons
Product Identifier
  • Calciferol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ergocalciferol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.