missing translation for 'onlineSavingsMsg'
Learn More

Calcifediol, MedChemExpress

Product Code. 16490498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490498 5 mg 5mg
16407816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490498 Supplier Medchem Express Supplier No. HY32351
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Calcifediol (25-hydroxy Vitamin D3), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.

Calcifediol (25-hydroxy Vitamin D3), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.

Specifications

Chemical Name or Material Calcifediol
CAS 19356-17-3
Color White
Purity Grade Notes Research
Molecular Formula C27H44O2
Quantity 5 mg
Synonym 25-hydroxy Vitamin D3
Solubility Information DMSO : ≥ 100 mg/mL (249.60 mM)
SMILES C=C(CC1)/C(C[C@H]1O)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CCCC(C)(C)O)CC3
Molecular Weight (g/mol) 400.64
Formula Weight 400.64
Percent Purity 98.93%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.