missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcein, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Calcein |
| CAS | 1461-15-0 |
| Molecular Formula | C30 H26 N2 O13 |
| Quantity | 1 g |
| Synonym | Fluorexon, Oftasceine, Fluorescein complexon, 2,7-Bis[N,N-bis(carboxymethyl)aminomethylene]fluorescein |
| InChI Formula | InChI=1S/C30H26N2O13/c33-21-7-23-19(5-15(21)9-31(11-25(35)36)12-26(37)38)30(18-4-2-1-3-17(18)29(43)45-30)20-6-16(22(34)8-24(20)44-23)10-32(13-27(39)40)14-28(41)42/h1-8,33-34H,9-14H2,(H,35,36)(H,37,38)(H,39,40)(H,41,42) |
| SMILES | OC(=O)CN(CC(=O)O)Cc1cc2c(Oc3cc(O)c(CN(CC(=O)O)CC(=O)O)cc3C24OC(=O)c5ccccc45)cc1O |
| IUPAC Name | 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 622.53 |
| Formula Weight | 622.1435 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction