missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Calcein AM
GREENER_CHOICE

Product Code. 18758218
Change view
Click to view available options
:
1mg
Product Code.
18758218 1mg
1 options
This item is not returnable. View return policy

Product Code. 18758218

Brand: Tocris Bioscience™ 5119/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell permeable compound; hydrolyzed to become fluorescent in living cells

Calcein AM is a cell permeable, non-fluorescent compound for monitoring cell viability, chemotaxis, cell adhesion and multidrug resistance; hydrolyzed by intracellular esterases to become fluorescent calcein in living cells. .
TRUSTED_SUSTAINABILITY

Specifications

CAS 890090-35-4
Quantity 1 mg
Product Type Calcein AM
Purity 95%
Molecular Weight (g/mol) M.W. (Observed): 994.86
Inhibitors Calcein AM

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.