missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CALCB Polyclonal antibody specifically detects CALCB in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | CALCB |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Alias | calcitonin-related polypeptide beta, FLJ30166 |
| Host Species | Mouse |
| Immunogen | CALCB (N/A, 1 a.a. - 127 a.a.) full-length human protein. MGFRKFSPFLALSILVLYQAGSLQAAPFRSALESSPDPATLSKEDARLLLAALVQDYVQMKASELKQEQETQGSSSAAQKRACNTATCVTHRLAGLLSRSGGMVKSNFVPTNVGSKAFGRRRRDLQA |
| Purification Method | Immunogen affinity purified |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?