missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DNase I, Bovine Pancreas
Shop All Merck Products
Click to view available options
Quantity:
25 mU
Description
- Native DNase I from bovine pancreas
- An endonuclease that cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position
Specifications
Specifications
| CAS | 9003-98-9 |
| Content And Storage | Store at −20°C |
| Formula Weight | 40000g/mol |
| MDL Number | MFCD00130918 |
| SMILES | * |
| IUPAC Name | Deoxyribonuclease |
| Molecular Weight (g/mol) | 0.00 |
| Quantity | 25 mU |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction