missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ DNase I, Bovine Pancreas

Product Code. 17797099 Shop All Merck Products
Change view
Click to view available options
Quantity:
25 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17797099 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17797099 Supplier MilliporeSigma™ Supplier No. 26091325MU
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Acts on single and double stranded DNA

  • Native DNase I from bovine pancreas
  • An endonuclease that cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position
TRUSTED_SUSTAINABILITY

Specifications

CAS 9003-98-9
Content And Storage Store at −20°C
Formula Weight 40000g/mol
Quantity 25 mU
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.