missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calbindin D28K Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143527
This item is not returnable.
View return policy
Description
Calbindin D28K Polyclonal Antibody for ICC/IF, IHC (P), IHC (PFA), Western Blot
Calbindin-D-28K (also termed vitamin D-dependent calcium-binding protein, or cholecalcin), is a highly conserved 28kDa calcium binding protein, with broad tissue distribution. It belongs, together with calmodulin, S-100, parvalbumin, troponin C and other proteins, to a family of low molecular weight calcium-binding proteins (CaBPs). These CaBPs have homologous primary structures, which contain conserved polypeptide folds of the EF-hand type for Ca2+ binding. Calbindin-D-28K is found predominantly in subpopulations of central and peripheral nervous system neurons, and in certain epithelial nervous system neurons, and in certain epithelial cells involved in Ca2+ transport such as distal tubular cells and cortical collecting tubules of the kidney, and in enteric neuroendocrine cells.
Specifications
| Calbindin D28K | |
| Polyclonal | |
| Unconjugated | |
| CALB1 | |
| Brain-2; CAB27; CaBP28K; Calb; CALB1; Calb-1; Calbindin; calbindin 1; calbindin 1, (28kD); calbindin 1, 28 kD; calbindin 1, 28kDa; calbindin D28; calbindin-28K; calbindin-D (28k); CalbindinD28K; CB; cerebellar Ca-binding protein spot 35 protein; cerebellar Ca-binding protein, spot 35 protein; D-28K; PCD-29; RTVL-H protein; spot 35 protein; Vitamin D-dependent calcium-binding protein, avian-type | |
| Chicken | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgY |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 0.02% sodium azide | |
| P04467, P05937, P07171, P12658 | |
| CALB1 | |
| Full-length recombinant human protein expressed in and purified from E. coli. | |
| RUO | |
| 12307, 539930, 793, 83839 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto