missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calbindin D28K Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143561
This item is not returnable.
View return policy
Description
Calbindin D28K Polyclonal Antibody for ICC/IF, IHC (P), IHC (PFA), Western Blot
Calbindin-D-28K (also termed vitamin D-dependent calcium-binding protein, or cholecalcin), is a highly conserved 28kDa calcium binding protein, with broad tissue distribution. It belongs, together with calmodulin, S-100, parvalbumin, troponin C and other proteins, to a family of low molecular weight calcium-binding proteins (CaBPs). These CaBPs have homologous primary structures, which contain conserved polypeptide folds of the EF-hand type for Ca2+ binding. Calbindin-D-28K is found predominantly in subpopulations of central and peripheral nervous system neurons, and in certain epithelial nervous system neurons, and in certain epithelial cells involved in Ca2+ transport such as distal tubular cells and cortical collecting tubules of the kidney, and in enteric neuroendocrine cells.
Specifications
| Calbindin D28K | |
| Polyclonal | |
| Unconjugated | |
| CALB1 | |
| Brain-2; CAB27; CaBP28K; Calb; CALB1; Calb-1; Calbindin; calbindin 1; calbindin 1, (28kD); calbindin 1, 28 kD; calbindin 1, 28kDa; calbindin D28; calbindin-28K; calbindin-D (28k); CalbindinD28K; CB; cerebellar Ca-binding protein spot 35 protein; cerebellar Ca-binding protein, spot 35 protein; D-28K; PCD-29; RTVL-H protein; spot 35 protein; Vitamin D-dependent calcium-binding protein, avian-type | |
| Chicken | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgY |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 0.02% sodium azide | |
| P04467, P05937, P07171, P12658 | |
| CALB1 | |
| Full-length recombinant human protein expressed in and purified from E. coli. | |
| RUO | |
| 12307, 539930, 793, 83839 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction