missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin-D28k Mouse anti-Human, Mouse, Rat, Clone: 1F8B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66394-1-IG-150UL
This item is not returnable.
View return policy
Description
Calbindin-D-28K (also termed vitamin D-dependent calcium-binding protein, or cholecalcin), is a highly conserved 28kDa calcium binding protein, with broad tissue distribution. It belongs, together with calmodulin, S-100, parvalbumin, troponin C and other proteins, to a family of low molecular weight calcium-binding proteins (CaBPs). These CaBPs have homologous primary structures, which contain conserved polypeptide folds of the EF-hand type for Ca2+ binding. Calbindin-D-28K is found predomitly in subpopulations of central and peripheral nervous system neurons, and in certain epithelial nervous system neurons, and in certain epithelial cells involved in Ca2+ transport such as distal tubular cells and cortical collecting tubules of the kidney, and in enteric neuroendocrine cells.Specifications
| Calbindin-D28k | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05937, P07171, P12658 | |
| Calb1 | |
| Calbindin-D28k Fusion Protein Ag5861 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1F8B9 | |
| Unconjugated | |
| CALB1 | |
| CAB27, CALB, CALB1, Calbindin, calbindin 1, 28kDa, Calbindin D28, D 28K | |
| Mouse | |
| Protein G | |
| RUO | |
| 12307, 793, 83839 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction