missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D/S100G Antibody (401025) [DyLight 680], Novus Biologicals™

Product Code. 30732976 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732976 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732976 Supplier Novus Biologicals Supplier No. FAB3320Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calbindin D/S100G Monoclonal antibody specifically detects Calbindin D/S100G in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D/S100G
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 401025
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CABP;CABP1;CABP9K;CALB3;Protein S100-G
Host Species Mouse
Immunogen E. coli-derived recombinant human Calbindin D, Met1-Asn261, Accession # P05937
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 795
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.