missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D-28K Mouse anti-Human, Clone: CALB1/3333, Novus Biologicals™

Mouse Monoclonal Antibody

£377.00

Specifications

Antigen Calbindin D-28K
Clone CALB1/3333
Concentration 0.2 mg/mL
Dilution ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18636087
View Documents
Novus Biologicals
NBP3-07364-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calbindin D-28K Monoclonal specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Calbindin D-28K
0.2 mg/mL
ELISA, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
10 mM PBS with 0.05% BSA
793
IgG2b κ
Protein A or G purified
CALB1/3333
ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Human
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.