missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D-28K Mouse anti-Human, Clone: CALB1/2782, Novus Biologicals™

Product Code. 18382426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382426 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382426 Supplier Novus Biologicals Supplier No. NBP30720820UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calbindin D-28K Monoclonal specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D-28K
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CALB1/2782
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Host Species Mouse
Immunogen Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 793
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.