missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Mouse anti-Human, Clone: CALB1/2364, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00
Specifications
| Antigen | Calbindin D-28K |
|---|---|
| Clone | CALB1/2364 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA, Protein Array |
| Applications | ELISA, Peptide Array |
Description
Calbindin D-28K Monoclonal specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Protein Array.Specifications
| Calbindin D-28K | |
| 0.2 mg/ml | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 793 | |
| IgG1 κ | |
| Protein A or G purified |
| CALB1/2364 | |
| ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
| Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title