missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody (rCALB1/8499), DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Calbindin D-28K |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rCALB1/8499 |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?