missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody (FGF2/7365), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24167DL594
439.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Calbindin D-28K | |
| Monoclonal | |
| DyLight 594 | |
| CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type | |
| Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 793 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| FGF2/7365 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction