missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D-28K Antibody (CL15060) [Unconjugated], Novus Biologicals™

Mouse Monoclonal Antibody

£321.00 - £459.00

Specifications

Antigen Calbindin D-28K
Clone CL15060
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:2500-1:5000, Immunohistochemistry-Paraffin 1:2500-1:5000
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30279339
View Documents Promotion Details
Novus Biologicals
NBP3-43645-100ul
100 μL
£459.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30279687
View Documents Promotion Details
Novus Biologicals
NBP3-43645-25ul
25 μL
£321.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

Calbindin D-28K
1 mg/mL
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
PBS, pH 7.2, 40% glycerol
793
IgG1
Protein A purified
CL15060
Western Blot 1 μg/mL, Immunohistochemistry 1:2500-1:5000, Immunohistochemistry-Paraffin 1:2500-1:5000
Monoclonal
Purified
RUO
Human, Mouse, Rat
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
This antibody was generated using a recombinant protein sequence of P05937, with the exact immunogen sequence remaining proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.