missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D-28K Antibody (4H7), Novus Biologicals™

Product Code. p-200056994 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18689036 0.025 mL 0.03mL
18661139 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18689036 Supplier Novus Biologicals Supplier No. NBP250048SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calbindin D-28K
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 4H7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry Free-Floating 1:1000
Formulation 50% PBS, 50% glycerol
Gene Accession No. P05937, P05937
Gene Alias CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Host Species Mouse
Immunogen Full-length recombinant human Calbindin D-28K. [UniProt# P05937]
Purification Method Immunogen affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 793
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.