Learn More
Beckman Coulter CALA (Calcium Arsenazo) Reagent, 4 x 15 mL
Brand: Beckman Coulter OSR60117
CALA (Calcium Arsenazo) Reagent, 4 x 15 mL
Safety and Handling
- Calcium Arsenazo
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Imidazole, 2,7-(bis(2-arsonophenylazo))-1,8-dihydroxynaphthalene-3,6-disulphonicacid, octylphenoxyp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.