missing translation for 'onlineSavingsMsg'
Learn More

CAL-130 Hydrochloride, MedChemExpress

Product Code. 16443047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443047

Brand: Medchem Express HY16122B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAL-130 is a PI3Kδ and PI3Kγ inhibitor with IC50s of 1.3 and 6.1 nM, respectively.

CAL-130 is a PI3Kδ and PI3Kγ inhibitor with IC50s of 1.3 and 6.1 nM, respectively.

Specifications

Chemical Name or Material CAL-130 Hydrochloride
Physical Form Solid
CAS 1431697-78-7
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H23ClN8O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (108.01 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1N(C2=CC=CC=C2C)C([C@H](C)NC3=C4N=CNC4=NC(N)=N3)=NC5=C1C(C)=CC=C5.[H]Cl
Molecular Weight (g/mol) 462.93
Formula Weight 462.93
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.