missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAGE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556271
This item is not returnable.
View return policy
Description
Immunogen sequence: DNNIENYSTN ALIQPVDTIS ISSLRQFETV CKFHWVEAFD DEMTEKPEFQ SQVYNYAKDN NIKQDSFKEE NPMETSVSAN TDQLGNEYF Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 45%.
CAGE1 (Cancer Antigen 1) is a Protein Coding gene. Diseases associated with CAGE1 include Fat Necrosis Of Breast and Amelogenesis Imperfecta, Type If. [GeneCards].
Specifications
| CAGE1 | |
| Polyclonal | |
| Unconjugated | |
| CAGE1 | |
| 4933427I01Rik; bA69L16.7; CAGE1; CAGE-1; cancer antigen 1; cancer/testis antigen 3; Cancer/testis antigen 3 homolog; cancer/testis antigen 95; cancer/testis antigen gene 1; cancer-associated gene 1 protein; Cancer-associated gene 1 protein homolog; CT3; CT3 homolog; CT95; Ctag3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC20 | |
| CAGE1 | |
| Recombinant protein corresponding to Human CAGE1. Recombinant protein control fragment (Product #RP-94920). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction