missing translation for 'onlineSavingsMsg'
Learn More

Caftaric acid, MedChemExpress

Product Code. 16481378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481378 5 mg 5mg
16458948 10 mg 10mg
16424817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481378 Supplier Medchem Express Supplier No. HYN0321
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Caftaric acid is a natural product.

Caftaric acid is a natural product.

Specifications

Chemical Name or Material Caftaric acid
CAS 67879-58-7
Color White
Purity Grade Notes Research
Molecular Formula C13H12O9
Quantity 5 mg
Synonym trans-Caftaric acid
Solubility Information H2O : 50 mg/mL (160.14 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (160.14 mM; Need ultrasonic)
SMILES O=C(O)[C@H](OC(/C=C/C1=CC=C(O)C(O)=C1)=O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 312.23
Formula Weight 312.23
Percent Purity 99.04%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.