missing translation for 'onlineSavingsMsg'
Learn More

Caficrestat, MedChemExpress

Product Code. 16490128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490128 5 mg 5mg
16447566 100 mg 100mg
16427668 10 mg 10mg
16423677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490128 Supplier Medchem Express Supplier No. HY18967
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Caficrestat (Aldose reductase-IN-1) is a inhibitor of aldose reductase with IC50 of 28.9 pM.

Caficrestat (Aldose reductase-IN-1) is a inhibitor of aldose reductase with IC50 of 28.9 pM.

Specifications

Chemical Name or Material Caficrestat
CAS 1355612-71-3
Color White
Purity Grade Notes Research
Molecular Formula C17H10F3N5O3S
Quantity 5 mg
Synonym Aldose reductase-IN-1 AT-001
Solubility Information DMSO : ≥ 28 mg/mL (66.45 mM)
SMILES O=C1C2=NC=CN=C2C(CC(O)=O)=NN1CC3=NC4=C(C=CC(C(F)(F)F)=C4)S3
Molecular Weight (g/mol) 421.35
Formula Weight 421.35
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.