missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, TRC

Product Code. 17981405
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981405 5 g 5g
17943125 1 g 1g
17931055 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981405 Supplier Toronto Research Chemicals Supplier No. TRCC0801005G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8 H10 N4 O2
Molecular Weight (g/mol) 194.19
Synonym Caffeine (8CI), 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 1,3,7-Trimethyl-2,6-dioxopurine, 1,3,7-Trimethyl-7H-purine-2,6-dione, 1,3,7-Trimethylxanthine, 7-Methyltheophylline, Alert-Pep, Asia migrine, Cafalgine, Cafeina, Caffedrine, Caffein, Cafipel, DHCplus, Dasin, Diurex, Durvitan, Guaranine, Koffein, Mateina, Methyltheobromine, Midron extra, Miudol, NSC 5036, New Cetamol, No-Doz, Palergot-C, Phensal, Refresh'n, Shape Plus, Stay Alert, Stim, Synalgos, Thein, Theine, Tri-Aqua, Vivarin, Wigraine
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
SMILES CN1C(=O)N(C)c2ncn(C)c2C1=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3
Formula Weight 194.0804
Percent Purity >95%
Chemical Name or Material Caffeine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.