missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, citrated

Product Code. p-7084042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15731779

Brand: Thermo Scientific Acros 461720250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Caffeine, citrated, Quantity: 100g, Melting Point: 159.0 deg.C to 161.0 deg.C, Molecular Weight: 386.31, CAS: 69-22-7, Formula Weight: 386.31, Health Hazard 1: GHS Signal Word: Warning, Health Hazard 2: GHS H Statement Harmful if swallowed. Causes serious eye irritation. | CAS: 69-22-7 | C14H18N4O9 | 386.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-22-7
Molecular Formula C14H18N4O9
Molecular Weight (g/mol) 386.32
MDL Number MFCD00044986
InChI Key RCQXSQPPHJPGOF-UHFFFAOYSA-N
Synonym caffeine citrate, caffeine, citrated, caffeina citrate, citrated caffein, caffeine citrate 1:1, caffeine citrated, unii-u26eo4675q, citric acid, compd. with caffeine 1:1, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, mixt. with 3,7-dihydro-1,3,7-trimethyl-1h-purine-2,6-dione, dsstox_cid_26938
PubChem CID 6241
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1,3,7-trimethylpurine-2,6-dione
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 159.0°C to 161.0°C
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 386.31
Chemical Name or Material Caffeine, citrated
compliance-icons
Product Identifier
  • Caffeine, citrated
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Caffeine, citrated

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.