missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, citrated

Product Code. 15741779 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15741779 100 g 100g
15731779 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15741779 Supplier Thermo Scientific Acros Supplier No. 461721000
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

Caffeine, citrated, Quantity: 100g, Melting Point: 159.0 deg.C to 161.0 deg.C, Molecular Weight: 386.31, CAS: 69-22-7, Formula Weight: 386.31, Health Hazard 1: GHS Signal Word: Warning, Health Hazard 2: GHS H Statement Harmful if swallowed. Causes serious eye irritation. | CAS: 69-22-7 | C14H18N4O9 | 386.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-22-7
Molecular Formula C14H18N4O9
Molecular Weight (g/mol) 386.32
MDL Number MFCD00044986
InChI Key RCQXSQPPHJPGOF-UHFFFAOYSA-N
Synonym caffeine citrate, caffeine, citrated, caffeina citrate, citrated caffein, caffeine citrate 1:1, caffeine citrated, unii-u26eo4675q, citric acid, compd. with caffeine 1:1, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, mixt. with 3,7-dihydro-1,3,7-trimethyl-1h-purine-2,6-dione, dsstox_cid_26938
PubChem CID 6241
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1,3,7-trimethylpurine-2,6-dione
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 159.0°C to 161.0°C
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 386.31
Chemical Name or Material Caffeine, citrated

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.