missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Citrate, TRC

Product Code. 30005891
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005891 25 g 25g
17920815 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005891 Supplier Toronto Research Chemicals Supplier No. TRCC08011525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-22-7
Molecular Formula C8 H10 N4 O2 . C6 H8 O7
Molecular Weight (g/mol) 386.31
Synonym 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, mixt. with 3,7-dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, Caffeine Citrate, Caffeine citrate (6CI), Caffeine, citrate (1:1) (7CI,8CI), 1H-Purine-2,6-dione, 3,7-dihydro-1,3,7-trimethyl-, mixt. contg. (9CI), Citrated caffeine
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1,3,7-trimethylpurine-2,6-dione
SMILES CN1C(=O)N(C)c2ncn(C)c2C1=O.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10N4O2.C6H8O7/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2;7-3(8)1-6(13,5(11)12)2-4(9)10/h4H,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 386.1074
Chemical Name or Material Caffeine Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.