missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Citrate, MP Biomedicals

Product Code. 11431346 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431346 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431346 Supplier MP Biomedicals Supplier No. 0520481880
  • ONLINE EXCLUSIVE PRICE
    £282.00
    £263.55 / 100g
    Save £18.45
    6% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 69-22-7
Molecular Formula C14H18N4O9
MDL Number MFCD00044986
Quantity 100 g
Synonym caffeine citrate, caffeine, citrated, caffeina citrate, citrated caffein, caffeine citrate 1:1, caffeine citrated, unii-u26eo4675q, citric acid, compd. with caffeine 1:1, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, mixt. with 3,7-dihydro-1,3,7-trimethyl-1h-purine-2,6-dione, dsstox_cid_26938
InChI Key RCQXSQPPHJPGOF-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1,3,7-trimethylpurine-2,6-dione
Molecular Weight (g/mol) 386.32
PubChem CID 6241
Formula Weight 386.32
Chemical Name or Material Caffeine Citrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.