missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Anhydrous MP Biomedicals

Product Code. 17832868 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 17832868

Brand: MP Biomedicals 0215011480

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Color White
Density 1.23g/mL
Quantity 100 g
Formula Weight 194.2
Physical Form Powder
Chemical Name or Material Caffeine Anhydrous, Anhydrous
compliance-icons
Product Identifier
  • Caffeine Anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Caffeine Anhydrous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.