missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine anhydrous, 99.7%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Specifications
Specifications
| Color | White |
| Density | 1.23g/mL |
| Assay Percent Range | 0.997 |
| Quantity | 250 g |
| Formula Weight | 194.2 |
| Physical Form | Powder |
| Chemical Name or Material | Caffeine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction