missing translation for 'onlineSavingsMsg'
Learn More

Caffeine anhydrous, 99.7%, MP Biomedicals™

Product Code. 11436015 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436015 500 g 500g
17812868 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436015 Supplier MP Biomedicals Supplier No. 0215011490
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Color White
Density 1.23g/mL
Assay Percent Range 0.997
Quantity 500 g
Formula Weight 194.2
Physical Form Powder
Chemical Name or Material Caffeine
compliance-icons
Product Identifier
  • Caffeine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.