missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 99%

Product Code. 11428643
Change view
Click to view available options
Quantity:
100 g
1000 g
Unit Size:
1000g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428643 1000 g 1000g
11364569 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428643 Supplier Thermo Scientific Alfa Aesar Supplier No. A10431.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 234°C to 236°C
Density 1.23
Assay Percent Range 99%
Quantity 1000 g
UN Number UN1544
Beilstein 17705
Merck Index 14,1636
Solubility Information Soluble in pyrrole,tetrahydrofuran+water mixture and ethyl acetate; Slightly soluble in water and ethanol.
IUPAC Name 1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material Caffeine
Show More Show Less
compliance-icons
Product Identifier
  • Caffeine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: caffeine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.