missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
70.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
10 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
- It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
- Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
Specifications
Acidity | 0.2mL of 0.01M NaOH max. |
Melting Point | 234.0°C to 239.0°C |
Color | White |
Density | 1.2300g/mL |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.1% max. (total impurities) (HPLC) |
Packaging | Plastic bottle |
Quantity | 500 g |
Beilstein | 26,461 |
Show More |
Safety and Handling
Product Identifier
- Caffeine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'