missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 98.5%, specified according to the req. of USP/BP

Product Code. p-81672 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
missing translation for 'unitSize'
10g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10254420

missing translation for 'mfr': Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC. | CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Acidity 0.2mL of 0.01M NaOH max.
Melting Point 234.0°C to 239.0°C
Color White
Density 1.2300g/mL
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. (total impurities) (HPLC)
Packaging Glass Bottle
Quantity 10 g
Beilstein 26,461
Merck Index 15,1639
Specific Gravity 1.23
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: 1g/66 mL alcohol,1g/50 mL acetone,freely soluble in pyrrole,soluble in ethyl acetate
Sublimation Point 178 °C
Residual Solvents meets the requirements
Formula Weight 194.19
Percent Purity 98.5%
Related Substances 0.5% max. (TLC)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Caffeine, 98.5%
Show More Show Less
compliance-icons
Product Identifier
  • Caffeine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.