missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeidine Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium 4-[N-Methyl-N-(N-methylaminocarbonyl)amino]-1-methylimidazole-5-carboxylate |
| Analyte or Component Names | Sodium 4-[N-Methyl-N-(N-methylaminocarbonyl)amino]-1-methylimidazole-5-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H11 N4 O3 . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sodium 4-[N-Methyl-N-(N-methylaminocarbonyl)-amino]-1-methylimidazole-5-carboxylate, Caffeidine acid sodium salt |
| InChI Formula | InChI=1S/C8H12N4O3.Na/c1-9-8(15)12(3)6-5(7(13)14)11(2)4-10-6;/h4H,1-3H3,(H,9,15)(H,13,14);/q;+1/p-1 |
| Solution Type | Neat |
| SMILES | [Na+].CNC(=O)N(C)c1ncn(C)c1C(=O)[O-] |
| IUPAC Name | sodium;3-methyl-5-[methyl(methylcarbamoyl)amino]imidazole-4-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?