missing translation for 'onlineSavingsMsg'
Learn More

Caffeic acid phenethyl ester, MedChemExpress

Product Code. 16499036
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499036 100 mg 100mg
16488928 10 mg 10mg
16471368 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499036 Supplier Medchem Express Supplier No. HYN0274
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Caffeic acid phenethyl ester is a NF-κB inhibitor.

Caffeic acid phenethyl ester is a NF-κB inhibitor.

Specifications

Chemical Name or Material Caffeic acid phenethyl ester
CAS 104594-70-9
Color White
Purity Grade Notes Research
Molecular Formula C17H16O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (351.73 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.52 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(OCCC1=CC=CC=C1)/C=C/C2=CC=C(O)C(O)=C2
Molecular Weight (g/mol) 284.31
Formula Weight 284.31
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.